Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128228.4 | NP_180239.1 | 1 - 2 | 528 | 112 | |
GenBank mRNA | AK175232.1 | - | 1 - 2 | 365 | 112 | |
AK229743.1 | - | 1 - 2 | 365 | 112 | ||
BT010564.1 | - | 1 - 2 | 291 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |