Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128239.1 | NP_180250.1 | 5 - 6 | 910 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |