Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128244.4 | NP_180255.1 | 2 - 3 | 962 | 78 | |
GenBank mRNA | AB084292.1 | - | 2 - 3 | 836 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |