Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128246.5 | NP_180257.3 | 19 - 20 | 7855 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |