Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDPMEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128250.4 | NP_180261.1 | 10 - 11 | 1116 | 97 | |
GenBank mRNA | AB037877.1 | - | 10 - 11 | 1060 | 97 | |
AX765854.1 | - | 10 - 11 | 1055 | 97 | ||
AY136394.1 | - | 10 - 11 | 1072 | 97 | ||
BT000208.1 | - | 10 - 11 | 1055 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |