Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDPMEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128250.4 | NP_180261.1 | 9 - 10 | 1025 | 100 | |
GenBank mRNA | AB037877.1 | - | 9 - 10 | 969 | 100 | |
AX765854.1 | - | 9 - 10 | 964 | 100 | ||
AY136394.1 | - | 9 - 10 | 981 | 100 | ||
BT000208.1 | - | 9 - 10 | 964 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |