Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RR13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336100.1 | NP_001318296.1 | 1 - 2 | 125 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons |