Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RR13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336100.1 | NP_001318296.1 | 6 - 7 | 1474 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |