Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128268.3 | NP_565635.1 | 9 - 10 | 1877 | 58 | |
GenBank mRNA | AF311221.1 | - | 9 - 10 | 1711 | 58 | |
AF428305.1 | - | 9 - 10 | 1716 | 58 | ||
AY039915.1 | - | 9 - 10 | 1723 | 58 | ||
AY142569.1 | - | 9 - 10 | 1687 | 58 | ||
AY143937.1 | - | 9 - 10 | 1687 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |