Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084499.2 | NP_001077968.1 | 4 - 5 | 311 | 73 | |
NM_001336108.1 | NP_001324367.1 | 4 - 5 | 336 | 73 | ||
NM_001336109.1 | NP_001324368.1 | 4 - 5 | 270 | 73 | ||
NM_128275.5 | NP_180285.4 | 4 - 5 | 351 | 73 | ||
GenBank mRNA | AK221283.1 | - | 4 - 5 | 289 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |