Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036357.1 | NP_001031434.1 | 4 - 5 | 555 | 98 | |
NM_128351.3 | NP_565663.1 | 4 - 5 | 711 | 98 | ||
NM_201822.1 | NP_973551.1 | 2 - 3 | 376 | 98 | ||
GenBank mRNA | AF412049.1 | - | 4 - 5 | 648 | 98 | |
BT003041.1 | - | 4 - 5 | 578 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |