Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036357.1 | NP_001031434.1 | 7 - 8 | 775 | 87 | |
NM_128351.3 | NP_565663.1 | 7 - 8 | 931 | 87 | ||
NM_201822.1 | NP_973551.1 | 5 - 6 | 596 | 87 | ||
GenBank mRNA | AF412049.1 | - | 7 - 8 | 868 | 87 | |
BT003041.1 | - | 7 - 8 | 798 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |