Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UXS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084503.2 | NP_001077972.1 | 4 - 5 | 420 | 102 | |
NM_001336187.1 | NP_001325413.1 | 4 - 5 | 517 | 102 | ||
NM_128436.5 | NP_180443.1 | 4 - 5 | 547 | 102 | ||
NM_201826.3 | NP_973555.1 | 4 - 5 | 620 | 102 | ||
GenBank mRNA | AY099703.1 | - | 4 - 5 | 435 | 102 | |
AY128899.1 | - | 3 - 4 | 351 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |