Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OEP16-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128449.3 | NP_180456.1 | 4 - 5 | 435 | 87 | |
GenBank mRNA | AY045593.1 | - | 4 - 5 | 352 | 87 | |
AY093782.1 | - | 4 - 5 | 233 | 87 | ||
DQ386642.1 | - | 4 - 5 | 233 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |