Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OEP16-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128449.3 | NP_180456.1 | 5 - 6 | 502 | 71 | |
GenBank mRNA | AY045593.1 | - | 5 - 6 | 419 | 71 | |
AY093782.1 | - | 5 - 6 | 300 | 71 | ||
DQ386642.1 | - | 5 - 6 | 300 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |