Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128494.5 | NP_180501.1 | 3 - 4 | 1141 | 78 | |
GenBank mRNA | AY080871.1 | - | 3 - 4 | 1116 | 78 | |
M93408.1 | - | 3 - 4 | 1068 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |