Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336227.1 | NP_001324174.1 | 15 - 16 | 2014 | 154 | |
NM_001336228.1 | NP_001324173.1 | 15 - 16 | 2116 | 154 | ||
NM_128543.4 | NP_029567.1 | 15 - 16 | 2050 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |