Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036367.2 | NP_001031444.2 | 16 - 17 | 2056 | 160 | |
NM_001202706.1 | NP_001189635.1 | 16 - 17 | 2056 | 160 | ||
NM_001336227.1 | NP_001324174.1 | 16 - 17 | 2033 | 154 | ||
NM_001336228.1 | NP_001324173.1 | 16 - 17 | 2135 | 154 | ||
NM_128543.4 | NP_029567.1 | 16 - 17 | 2069 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |