Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035662.2 | NP_001030739.1 | 4 - 5 | 787 | 87 | |
NM_001338505.1 | NP_001319605.1 | 3 - 4 | 960 | 87 | ||
NM_113005.4 | NP_188748.1 | 4 - 5 | 863 | 87 | ||
GenBank mRNA | BT002734.1 | - | 3 - 4 | 571 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |