Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035662.2 | NP_001030739.1 | 6 - 7 | 973 | 116 | |
NM_001338505.1 | NP_001319605.1 | 5 - 6 | 1146 | 116 | ||
NM_113005.4 | NP_188748.1 | 6 - 7 | 1049 | 116 | ||
GenBank mRNA | BT002734.1 | - | 5 - 6 | 757 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |