Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203015.2 | NP_001189944.1 | 11 - 12 | 4648 | 118 | |
NM_001338517.1 | NP_001326627.1 | 12 - 13 | 4559 | 118 | ||
NM_001338518.1 | NP_001326629.1 | 11 - 12 | 4538 | 118 | ||
NM_001338519.1 | NP_001326630.1 | 11 - 12 | 4386 | 118 | ||
NM_001338520.1 | NP_001326628.1 | 12 - 13 | 4330 | 118 | ||
NM_113020.3 | NP_188762.3 | 11 - 12 | 4496 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |