Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338521.1 | NP_001325430.1 | 12 - 13 | 1146 | 71 | |
NM_113023.4 | NP_566680.2 | 13 - 14 | 1399 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |