Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113067.4 | NP_188809.2 | 2 - 3 | 616 | 94 | |
GenBank mRNA | AK226403.1 | - | 2 - 3 | 510 | 94 | |
AK316978.1 | - | 2 - 3 | 508 | 94 | ||
AY140000.1 | - | 2 - 3 | 508 | 94 | ||
BT010397.1 | - | 2 - 3 | 436 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |