Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT71B8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113075.3 | NP_188817.1 | 1 | 1643 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |