Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113082.4 | NP_566696.2 | 2 - 3 | 661 | 85 | |
GenBank mRNA | AY063961.1 | - | 2 - 3 | 486 | 85 | |
AY096402.1 | - | 2 - 3 | 375 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |