Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113082.4 | NP_566696.2 | 4 - 5 | 769 | 150 | |
GenBank mRNA | AY063961.1 | - | 4 - 5 | 594 | 150 | |
AY096402.1 | - | 4 - 5 | 483 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |