Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113175.2 | NP_566717.1 | 1 - 2 | 258 | 138 | |
GenBank mRNA | AY099653.1 | - | 1 - 2 | 129 | 138 | |
AY128837.1 | - | 1 - 2 | 63 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |