Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338596.1 | NP_001325669.1 | 2 - 3 | 396 | 94 | |
NM_113175.2 | NP_566717.1 | 2 - 3 | 408 | 94 | ||
GenBank mRNA | AY099653.1 | - | 2 - 3 | 279 | 94 | |
AY128837.1 | - | 2 - 3 | 213 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |