Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKP-ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113196.3 | NP_566720.1 | 5 - 6 | 1676 | 114 | |
GenBank mRNA | AY056793.1 | - | 5 - 6 | 1600 | 114 | |
AY058084.1 | - | 5 - 6 | 1627 | 114 | ||
BT002329.1 | - | 5 - 6 | 1492 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |