Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035670.1 | NP_001030747.1 | 1 - 2 | 628 | 99 | |
NM_113226.5 | NP_188966.1 | 2 - 3 | 550 | 99 | ||
GenBank mRNA | AK176617.1 | - | 2 - 3 | 550 | 99 | |
AK176693.1 | - | 2 - 3 | 550 | 99 | ||
AY550296.1 | - | 2 - 3 | 267 | 99 | ||
BT026046.1 | - | 2 - 3 | 267 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |