Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KINESIN-12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035673.2 | NP_001030750.1 | 11 - 12 | 1489 | 115 | |
NM_113271.4 | NP_189009.2 | 11 - 12 | 1519 | 115 | ||
GenBank mRNA | AF540489.1 | - | 11 - 12 | 1491 | 115 | |
AY124006.1 | - | 11 - 12 | 1487 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |