Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KINESIN-12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035673.2 | NP_001030750.1 | 1 - 2 | 413 | 75 | |
NM_113271.4 | NP_189009.2 | 1 - 2 | 443 | 75 | ||
GenBank mRNA | AF540489.1 | - | 1 - 2 | 415 | 75 | |
AY124006.1 | - | 1 - 2 | 411 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |