Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KINESIN-12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035673.2 | NP_001030750.1 | 2 - 3 | 556 | 95 | |
NM_113271.4 | NP_189009.2 | 2 - 3 | 586 | 95 | ||
GenBank mRNA | AF540489.1 | - | 2 - 3 | 558 | 95 | |
AY124006.1 | - | 2 - 3 | 554 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |