Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338655.1 | NP_001327897.1 | 3 - 4 | 711 | 65 | |
NM_113284.3 | NP_189022.1 | 4 - 5 | 1035 | 65 | ||
GenBank mRNA | AK117618.1 | - | 4 - 5 | 827 | 65 | |
BT005102.1 | - | 4 - 5 | 780 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |