Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113319.5 | NP_189056.2 | 5 - 6 | 1400 | 111 | |
GenBank mRNA | AK221324.1 | - | 5 - 6 | 1401 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |