Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AtSec20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113338.5 | NP_566746.1 | 6 - 7 | 687 | 103 | |
GenBank mRNA | BT025252.1 | - | 5 - 6 | 525 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |