Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AtSec20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113338.5 | NP_566746.1 | 7 - 8 | 803 | 110 | |
GenBank mRNA | BT025252.1 | - | 6 - 7 | 641 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |