Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203032.1 | NP_001189961.1 | 4 - 5 | 840 | 80 | |
NM_113342.4 | NP_189078.2 | 3 - 4 | 864 | 80 | ||
GenBank mRNA | BT004300.1 | - | 3 - 4 | 764 | 80 | |
BT008604.1 | - | 3 - 4 | 673 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |