Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VRN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338682.1 | NP_001327228.1 | 1 - 2 | 667 | 61 | |
NM_113351.3 | NP_189087.1 | 1 - 2 | 734 | 61 | ||
GenBank mRNA | EF064791.1 | - | 1 - 2 | 424 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |