Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH2C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113359.4 | NP_566749.1 | 8 - 9 | 1498 | 65 | |
GenBank mRNA | AF349448.1 | - | 8 - 9 | 1263 | 65 | |
AY056398.1 | - | 8 - 9 | 1269 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |