Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338693.1 | NP_001319632.1 | 1 - 2 | 261 | 107 | |
NM_113367.3 | NP_566753.1 | 4 - 5 | 830 | 107 | ||
NM_202628.2 | NP_974357.1 | 4 - 5 | 820 | 107 | ||
GenBank mRNA | AY040011.1 | - | 4 - 5 | 829 | 107 | |
BT033138.1 | - | 1 - 2 | 190 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |