Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPGEF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113373.2 | NP_189105.1 | 1 - 2 | 354 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |