Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XTH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113409.4 | NP_189141.1 | 2 - 3 | 322 | 90 | |
GenBank mRNA | DQ446697.1 | - | 2 - 3 | 301 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |