Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP15-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113428.3 | NP_566762.1 | 4 - 5 | 339 | 78 | |
GenBank mRNA | AY062776.1 | - | 4 - 5 | 292 | 78 | |
AY128760.1 | - | 4 - 5 | 264 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |