Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338744.1 | NP_001319641.1 | 5 - 6 | 586 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |