Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338744.1 | NP_001319641.1 | 7 - 8 | 720 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |