Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338755.1 | NP_001319643.1 | 1 - 2 | 455 | 91 | |
NM_001338756.1 | NP_001327930.1 | 2 - 3 | 459 | 91 | ||
NM_113450.5 | NP_566769.1 | 2 - 3 | 459 | 91 | ||
GenBank mRNA | AF198179.1 | - | 1 - 2 | 184 | 91 | |
AF360152.1 | - | 1 - 2 | 290 | 91 | ||
AY142597.1 | - | 1 - 2 | 134 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |