Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338755.1 | NP_001319643.1 | 2 - 3 | 700 | 88 | |
NM_001338756.1 | NP_001327930.1 | 3 - 4 | 704 | 88 | ||
NM_113450.5 | NP_566769.1 | 3 - 4 | 704 | 88 | ||
GenBank mRNA | AF198179.1 | - | 2 - 3 | 429 | 88 | |
AF360152.1 | - | 2 - 3 | 535 | 88 | ||
AY142597.1 | - | 2 - 3 | 379 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |