Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203045.1 | NP_001189974.1 | 9 - 10 | 3097 | 73 | |
NM_001203046.1 | NP_001189975.1 | 9 - 10 | 2674 | 73 | ||
NM_001338761.1 | NP_001327702.1 | 10 - 11 | 3282 | 73 | ||
NM_001338762.1 | NP_001327703.1 | 10 - 11 | 3121 | 73 | ||
NM_001338763.1 | NP_001327701.1 | 9 - 10 | 4097 | 73 | ||
NM_113468.5 | NP_189197.2 | 8 - 9 | 3680 | 73 | ||
GenBank mRNA | AB087408.1 | - | 8 - 9 | 3112 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |