Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113489.3 | NP_189215.1 | 5 - 6 | 1539 | 101 | |
GenBank mRNA | AF066079.1 | - | 5 - 6 | 1398 | 101 | |
AY037262.1 | - | 5 - 6 | 1412 | 101 | ||
BT002343.1 | - | 5 - 6 | 1340 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |